missing translation for 'onlineSavingsMsg'
Learn More

Cerium sputtering target, 76.2mm dia x 3.18mm thick, Thermo Scientific Chemicals

Catalog Number 15452577
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 Ea.
CAS 7440-45-1
Odor Odorless
Molecular Formula Ce
MDL Number MFCD00010924
UN Number UN1333
Synonym cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium
InChI Key GWXLDORMOJMVQZ-UHFFFAOYSA-N
SMILES [Ce]
IUPAC Name cerium
Molecular Weight (g/mol) 140.12
PubChem CID 23974
ChEBI CHEBI:33369
Chemical Name or Material Cerium sputtering target, 76.2mm dia. x 3.18mm thick
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'