missing translation for 'onlineSavingsMsg'
Learn More

Cerium powder, -60 mesh, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 15432557
Click to view available options
Quantity:
5 g
25 g
7440-45-1
Ce
140.12
MFCD00010924
GWXLDORMOJMVQZ-UHFFFAOYSA-N
cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium
23974
CHEBI:33369
cerium
[Ce]
7440-45-1
Ce
140.12
MFCD00010924
GWXLDORMOJMVQZ-UHFFFAOYSA-N
cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium
23974
CHEBI:33369
cerium
[Ce]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Color Silver
UN Number UN3078
Quantity 5 g
Percent Purity 99.9%
Mesh Size −200 Mesh
Odor Odorless
Sensitivity Air and moisture sensitive
Form Powder
Assay (REO)
Chemical Name or Material Cerium powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'