Learn More
Cerium IV Sulphate 0.1 M (0.1 N) Analytical Volumetric Solution (AVL), Reagecon
Fully factorized, high purity, certified, stable product, developed and tested for oxidation/reduction titrations.
Brand: Reagecon CS2011
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 2796 Chem class : 8 Pack group : II
Description
- Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
- Accurate, stable, reproducible and fully factorized.
- Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
- Stability is proven (either opened or unopened) for the shelf life of this product.
- Certified to a specification of ± 0.1%.
- Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
- May be used for manual titrations, or automatic titrations, to determine the end points by detection of any of several properties.
- Oxidation/reduction titrations depend on changes in the electropotential of the oxidation reduction system and always involves a reaction between an oxidizing and reducing pair.
Specifications
Accuracy ± 0.01% | |
HDPE Bottle GL45 | |
Titration |
0.1 M (0.1 N) | |
1 L | |
Liquid |
Safety and Handling
- CERIUM SULPHATE 0.10M (0.10N)
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Cerium (IV) SulfateTetrahydrate, sulphuric acid