missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) and Dilute Nitric acid, Etchant Solution, Ce(IV) concentration 0.25N, Thermo Scientific Chemicals

Catalog Number p-7051849
Click to view available options
Quantity:
1 L
4 L
This item is not returnable. View return policy
MFCD01866052
This item is not returnable. View return policy
MFCD01866052

It is widely applied as etchant solution in industrial purpose to chemically remove layers from the surface of a wafer during manufacturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Quantity 1 L
UN Number UN3098
Solubility Information Soluble in water
Concentration or Composition (by Analyte or Components) Ce(IV) concentration 0.25N
Chemical Name or Material Cerium(IV) and Dilute Nitric acid, Etchant Solution

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'