missing translation for 'onlineSavingsMsg'
Learn More

Cerium ingot, 99% (REO), Thermo Scientific Chemicals

Catalog Number p-7059107
Click to view available options
Quantity:
250 g
1 kg
7440-45-1
Ce
140.12
MFCD00010924
GWXLDORMOJMVQZ-UHFFFAOYSA-N
cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium
23974
CHEBI:33369
cerium
[Ce]
7440-45-1
Ce
140.12
MFCD00010924
GWXLDORMOJMVQZ-UHFFFAOYSA-N
cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium
23974
CHEBI:33369
cerium
[Ce]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under mineral oil
TRUSTED_SUSTAINABILITY
Color Gray
UN Number UN1333
Quantity 250 g
Percent Purity 99%
Odor Odorless
Assay (REO)
Chemical Name or Material Cerium ingot

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'