missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4407354
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
12014-93-6
Ce2S3
376.41
MFCD00016007
MMXSKTNPRXHINM-UHFFFAOYSA-N
cerium sulfide, dicerium trisulphide, ceric sulfide, cerium 3+ trisulfide, dicerium 3+ trisulfide, dicerium 3+ trisulfanediide
159394
[S--].[S--].[S--].[Ce+3].[Ce+3]
This item is not returnable. View return policy
12014-93-6
Ce2S3
376.41
MFCD00016007
MMXSKTNPRXHINM-UHFFFAOYSA-N
cerium sulfide, dicerium trisulphide, ceric sulfide, cerium 3+ trisulfide, dicerium 3+ trisulfide, dicerium 3+ trisulfanediide
159394
[S--].[S--].[S--].[Ce+3].[Ce+3]

Used as pharmaceutical intermediates and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 2100°C
Color Red to Brown
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information Insoluble in water.
IUPAC Name cerium(3+);trisulfide
Formula Weight 376.43
Odor Rotten eggs
Appearance Red-brown powder
Density 5.02 g/cm3 at 20°C
Chemical Name or Material Cerium(III) sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'