missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) oxalate hydrate, REacton™, 99.989% (REO), Thermo Scientific™

Catalog Number p-7049211
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
15750-47-7
C6Ce2O12
544.29
MFCD00013082
ZMZNLKYXLARXFY-UHFFFAOYSA-H
cerium iii oxalate hydrate, cerium iii oxalatehydrate, cerium iii oxalate decahydrate
91998046
[Ce+3].[Ce+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
This item is not returnable. View return policy
15750-47-7
C6Ce2O12
544.29
MFCD00013082
ZMZNLKYXLARXFY-UHFFFAOYSA-H
cerium iii oxalate hydrate, cerium iii oxalatehydrate, cerium iii oxalate decahydrate
91998046
[Ce+3].[Ce+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point (decomposition)
Physical Form Crystalline
Quantity 10 g
UN Number UN3288
Merck Index 14,2001
IUPAC Name cerium;oxalic acid;hydrate
Formula Weight 544.29 (Anhydrous)
Percent Purity 99.989%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Cerium(III) oxalate hydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'