missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) chloride, 99.9%, pure, anhydrous, Thermo Scientific Chemicals

Catalog Number p-3734717
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7790-86-5
CeCl3
246.48
MFCD00010929
VYLVYHXQOHJDJL-UHFFFAOYSA-K
cerium iii chloride, cerous chloride, cerium iii chloride, anhydrous, cerium-144 chloride, cerium chloride generic, cerium-chloride, cerium chloride octahydrate, cerous iii-chloride
24636
CHEBI:35458
cerium(3+);trichloride
[Cl-].[Cl-].[Cl-].[Ce+3]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7790-86-5
CeCl3
246.48
MFCD00010929
VYLVYHXQOHJDJL-UHFFFAOYSA-K
cerium iii chloride, cerous chloride, cerium iii chloride, anhydrous, cerium-144 chloride, cerium chloride generic, cerium-chloride, cerium chloride octahydrate, cerous iii-chloride
24636
CHEBI:35458
cerium(3+);trichloride
[Cl-].[Cl-].[Cl-].[Ce+3]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1727.0°C
Melting Point 848.0°C
Color White to Gray
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.90%
Linear Formula CeCl3
Merck Index 15, 1996
Solubility Information Solubility in water: soluble
Formula Weight 246.48
Percent Purity 99.9%
Grade Pure
Packaging Glass bottle
Chemical Name or Material Cerium(III) chloride, Anhdrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'