missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerium(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals
31.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Cerium(III) bromide is used as an additive for chemical reagent. Undoped single crystals of CeBr3 have shown promise as a γ-ray scintillation detector in nuclear non-proliferation testing, medical imaging, environmental remediation, and oil exploration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Boiling Point | 1560°C |
Melting Point | 733°C |
Color | Gray to White |
Physical Form | Powder |
Quantity | 1 g |
Merck Index | 14,1995 |
Solubility Information | Soluble in water. |
Formula Weight | 379.83 |
Percent Purity | 99.9% |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'