Learn More
Cerium(III) 2-ethylhexanoate, 49% in 2-ethylhexanoic acid, Ce 12%, Thermo Scientific Chemicals
Description
Used as catalyst, paint dryer, plastic Additive. As the the precursor in preparation of inorganic nanoparticles in oil-in-water microemulsions. The Toluene-based precursor solution was composed by mixing gadolinium (III) 2-ethylhexanoate and cerium (III) 2-ethylhexanoate to achieve the right stoichiometry. For the ceria/zirconia precursor, cerium(III) 2-ethylhexanoate and zirconium(IV) 2-ethylhexanoate were mixed to result in a metal molar ratio Ce/Zr of 1:1 and diluted with xylene to a total metal. Thin films of CeO 2 were prepared by a photochemical method using thin amorphous films of cerium(III) 2-ethylhexanoate as the precursor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 1.08 g/mL |
Flash Point | >110°C (230°F) |
Quantity | 500 g |
Solubility Information | Insoluble in water. |
Formula Weight | 569.74 |
Concentration or Composition (by Analyte or Components) | 49% in 2-ethylhexanoic acid, Ce 12% |
Physical Form | Viscous Liquid |
Chemical Name or Material | Cerium(III) 2-ethylhexanoate |
RUO – Research Use Only