missing translation for 'onlineSavingsMsg'
Learn More

Ceramide, Tocris Bioscience™

Ser/Thr protein phosphatase activator

Brand:  Tocris (Bio-Techne) 0744/50

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13437943

  • £393.92 / 50mg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A potent modulator of cell proliferation and differentiation. Activates protein phosphatase-1 (PP1) and -2A (PP2A), as well as ceramide-activated protein phosphatase (CAPP) in vitro.
Specifications

Chemical Identifiers

3102-57-6
341.536
n-acetylsphingosine, c2-ceramide, c2 ceramide, n-acetyl-sphing-4-enine, n-acetyl-ceramide, d-erythro-sphingosine, n-acetyl, n-acetyl-d-sphingosine, acetyl ceramide, c2 ceramide d18:1/2:0, acetamide, n-2-hydroxy-1-hydroxymethyl-3-heptadecenyl-, r-r*,s*-e
CHEBI:46979
CCCCCCCCCCCCCC=CC(C(CO)NC(=O)C)O
C20H39NO3
BLTCBVOJNNKFKC-QUDYQQOWSA-N
5497136
N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]acetamide

Specifications

Ceramide
3102-57-6
n-acetylsphingosine, c2-ceramide, c2 ceramide, n-acetyl-sphing-4-enine, n-acetyl-ceramide, d-erythro-sphingosine, n-acetyl, n-acetyl-d-sphingosine, acetyl ceramide, c2 ceramide d18:1/2:0, acetamide, n-2-hydroxy-1-hydroxymethyl-3-heptadecenyl-, r-r*,s*-e
CCCCCCCCCCCCCC=CC(C(CO)NC(=O)C)O
341.536
CHEBI:46979
>98%
50mg
C20H39NO3
BLTCBVOJNNKFKC-QUDYQQOWSA-N
N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]acetamide
5497136
341.53
Videos