Learn More
CEP55 Mouse anti-Human, Clone: EMRC10-11-55, eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-9809-82
Code : Z2
Additional Details : Weight : 0.01000kg
Description
Description: The monoclonal antibody EMRC10-11-55 recognizes the human centrosomal protein (Cep55). Cep55 is a 55 kDa microtubule-bundling protein that is found throughout the cell but specifically localizes to the mitotic spindle during metaphase and the
Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered. Plays a role in mitotic exit and cytokinesis. Not required for microtubule nucleation. Recruits PDCD6IP and TSG101 to midbody during cytokinesis.Specifications
CEP55 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q53EZ4 | |
CEP55 | |
Affinity chromatography | |
RUO | |
55165 | |
4° C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
EMRC10-11-55 | |
Unconjugated | |
CEP55 | |
C10orf3, CT111, URCC6, cancer/testis antigen 111, centrosomal protein of 55 kDa, up-regulated in colon cancer 6 | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only.