Learn More
Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Type 11302, 3μm
Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Type 11302, 3μm are recommended for a broad range of applications including liquid filtering for sample preparation.
Brand: Sartorius Stedim Biotech™ 11302--90------G
Additional Details : Weight : 0.06400kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Description
- Cellulose nitrate (CN), mixed cellulose ester (MCE), membrane filters are composed of cellulose nitrate and a small content of cellulose acetate.
- Recommended for a broad range of applications such as the filtration of aqueous solutions or sample preparation.
- Also suitable for air/gas filtration, clarification, particle analysis
- Sterilizable by autoclave, gamma irradiation, or ETO
- Non-sterile, bulk-packed
Specifications
White | |
3 μm | |
11302 | |
25/pk. | |
90 mm |
Cellulose Nitrate, Mixed Cellulose Ester | |
Non-sterile | |
Hydrophilic | |
130 μm |
Safety and Handling
- Danger
Hazard Category
- Flammable solid Category 1
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P230-Keep wetted with .
- P233-Keep container tightly closed.
- P234-Keep only in original container.
- P243-Take precautionary measures against static discharge.
- P370+P378-In case of fire: Use to extinguish.
- P372-Explosion risk in case of fire.
- P375-Fight fire remotely due to the risk of explosion.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.