missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cellulose microcrystalline, extra pure, average particle size 90μm, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
Specifications
Color | White |
pH | 5 to 7 |
Physical Form | Fine Crystalline Powder or Granular Powder |
Linear Formula | (C12H20O10)n |
Solubility Information | Solubility in water: insoluble. |
Quantity | 25 kg |
Grade | Extra Pure |
Infrared Spectrum | Authentic |
Loss on Drying | 1.5% max. |
Packaging | Fiber Drum |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'