missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cellulose, microcrystalline

Catalog Number p-3346690
56.15 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
10,000 g
2500 g
500 g
9004-34-6
MFCD00081512
9004-34-6
MFCD00081512

Cellulose, microcrystalline is used as an emulsifier, bulking agent in food production, extender and anti-caking agent. It is also used in vitamin supplements or tablets. It acts as an alternative to carboxymethylcellulose. It finds application in partition chromatography. Further, it is used as a thickener and stabilizer. In addition to this, it is used as an excipient in the pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Microcrystalline
Merck Index 14,1965
Solubility Information Slightly soluble in sodium hydroxide. Insoluble in water,ethanol,ether and dilute mineral acids.
Quantity 500 g
Chemical Name or Material Cellulose, microcrystalline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'