missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cellulose acetate, average M.W. 100,000

Catalog Number p-84122
Click to view available options
Quantity:
25 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
9004-35-7
(C6H7O5)nC2H3O
MFCD00081496
This item is not returnable. View return policy
9004-35-7
(C6H7O5)nC2H3O
MFCD00081496

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Powder
Merck Index 14, 1966
Quantity 25 g
Formula Weight 100,000 (Average )
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Plastic bottle
Chemical Name or Material Cellulose acetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'