missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Cellulase (T. reesei)
Used for Enzyme
Brand: Abnova™ P5235.1g
Additional Details : Weight : 0.00010kg
Description
#REF!Specifications
Enzyme Activity | |
Native protein purified from Trichoderma reesei ATCC#26921 | |
Store at 4°C. | |
CELLULASE | |
≥25 units per mg dry weight. One Unit releases 0.01 milligrams of glucose per hour from microcrystalline cellulose at 37°C, pH 5.0. | |
None | |
Lyophilized |
Cellulase (T. reesei) | |
1 g | |
RUO | |
Cellulase Assay | |
Fungi | |
Native protein purified from Trichoderma reesei ATCC#26921 |