Learn More
Biodesign™ CellMicroSieves™
£28.15 - £134.00
Product Code | Brand | Type | Pore Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Pore Size | Price | Quantity & Availability | ||||
12924277
|
Biodesign™
N100C |
Circle | 100 um |
£34.60
Pack of 10 |
Estimated Shipment: 01-07-2024
Log in to see stock. |
||||
12974277
|
Biodesign™
N20C |
Circle | 20 um |
£28.15
Pack of 10 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
12964267
|
Biodesign™
N20R |
Roll | 20 um |
£134.00
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
12964277
|
Biodesign™
N25C |
Circle | 25 um |
£49.90
Pack of 10 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
12944277
|
Biodesign™
N50C |
Circle | 50 um |
£38.00
Pack of 10 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
||||
12934277
|
Biodesign™
N70C |
Circle | 70 um |
£28.15
Pack of 10 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
||||
12924267
|
Biodesign™
N70R |
Roll | 70 um |
£99.85
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
||||
Description
Unlike traditional filters, CellMicroSieves™ are a precision woven matrix of pure nylon filaments that is available with pores ranging from 5μL to 200μL. CellMicroSieves™ do not wet and contains no dyes, detergents or extractables. Extremely low protein affinity means that cells and proteins will not bind to CellMicroSieves™. They have a wide variety of distinctive uses a specific sized porous screens in the laboratory. AccessoriesPolypropylene sieve holders (2 piece) holders with adjustable metal tightener or filtration units, type A borosilicate glass with stainless steel filter support and aluminum clamp or type B complete polysulfone filtration unit with polypropylene filter support are available. Both filtration units have a 300mL capacity are autoclavable and reusable. Use sheets and rolls with sieve holders and circles with filtration units to hold securely when processing samples.- A pure nylon product that is biologically inert, resistant to most acidic or basic conditions and many solvents
- Autoclavable and can be repeatedly washed and reused
- Rapid collection of tissue culture cells without centrifugation
- Isolation of specific cell types from a dissociated tissue sample, allowing only a single type of cell to pass through
- Separation of different sized cells from a mixture of cell types
- Concentration and isolation of cells from a bioreactor
- Prefilter to prevent clogging of final filter CellMicroSieves™ allow the rapid gentle isolation of cells, organelles or cellular debris.