missing translation for 'onlineSavingsMsg'
Learn More

Corning™ cellgro™ DMEM Liquid Classical Cell Culture Media

Products undergo a variety of tests to determine functionality and toxicity using growth-promotion analysis. Corning™ cellgro™ DMEM Liquid Classical Cell Culture Media support cell culture growth and subsititution of Pyridoxine HCl for Pyridoxal ensures improved stability over original formulation.

Brand:  Corning™ 10-102-CVR

Code : NEW

Additional Details : Weight : 3.76000kg

 View more versions of this product

Product Code. 15333611

  • £107.16 / Pack of 6


Estimated Shipment date
from Supplier 23-02-2018
Add to basket

Description

Description

  • Modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
  • Original formulation was used to culture embryonic mouse cells
  • Has been modified to support variety of normal and transformed cells, such as primary cultures of mouse and chicken cells
  • Formulations can differ in amount or absence of components such as glucose, sodium pyruvate, sodium bicarbonate and L-glutamine
  • Tested to determine pH and osmolality
  • Low levels of endotoxin, and absence of contamination
  • Tested for bacterial endotoxin using the Limulus Amoebocyte Lysate (LAL) Chromogenic Assay
  • Osmolality: 335±mOsm/kg H2O
  • Mycoplasma: Tested Negative
  • Sterility: Pass
Specifications

Specifications

1X
To assess growth promotion and cytotoxicity characteristics
Inorganic Salts: 200mg/L CaCl2 (anhydrous), 0.1mg/L Fe(NO3)3.9H2O, 400mg/L KCl, 97.7mg/L MgSO4 (anhydrous), 6400mg/L NaCl, 125mg/L NaH2PO4.H2O, 3700mg/L NaHCO3; Amino Acids: 84mg/L L-Arginine. HCl, 62.57mg/L L-Cystine.2HCl, 869mg/L L-Aianyi-L-glutamine, 30mg/L Glycine, 42mg/L L-Histidine.HCl.H2O, 104.8mg/L L-Isoleucine, 104.8mg/L L-Leucine, 146.2mg/L L-Lysine. HCl, 30mg/L L-Methionine, 66mg/L L-Phenylalanine, 42mg/L L-Serine, 95.2mg/L L-Threonine, 16mg/L L-Tryptophan, 103.79mg/L L-Tyrosine.2Na.2H2O, 20mg/L L-Valine; Vitamins: 4mg/L D-Calcium pantothenate, 4mg/L Choline chloride, 4mg/L Folic Acid, 7.2mg/L i-Inositol, 4mg/L Nicotinamide, 4mg/L Pyridoxine. HCl, 0.4mg/L Riboflavin, 4mg/L Thiamine. HCl; Other: 4500mg/L D-Glucose, 15mg/L Phenol red. Na.
6 x 500mL
Sodium Pyruvate
<0.25EU/mL
Liquid
DMEM Media
Corning glutagro&trade supplement, 4.5g/L Glucose, Phenol red
2 to 8°C
Videos