missing translation for 'onlineSavingsMsg'
Learn More

Biochrom™ Cell Holder

Brand:  Biochrom™ 80-2106-05

Code : GX

Additional Details : Weight : 0.28000kg

 View more versions of this product

Product Code. 10680951

  • £163.00 / Each


Estimated Shipment
27-07-2018
Add to basket

If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.

 

Description

Description

An adjustable single cell holder for 10-40mm pathlength cells. Operational temperature range from ambient to 100°C and compatible with Ultrospec 2100 pro, 3100, 3300, 4300, 5300 and 6300 spectrophotometer models. Suitable for enzyme kinetics studies that need to be under thermostatic control from a circulation bath. This water-heated cell holder is an alternative to the Peltier heated versions.
  • Adjustable single cell holder for 10 to 40mm pathlength cells.
  • Temperature range from ambient to 100°C.
  • Compatible with Ultrospec 2100 pro, 3100, 3300, 4300, 5300, and 6300 models.
Specifications

Specifications

Ultrospec 2100 pro, 3100, 3300, 4300, 5300, and 6300 spectrophotometers
Ultrospec 2100 pro, 3100, 3300, 4300, 5300, and 6300 spectrophotometers
Cell Holder
Videos