missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cefuroxime sodium salt, 95%, Thermo Scientific™
Click to view available options
Quantity:
5g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Merck Index | 14,1954 |
Solubility Information | Freely soluble in water |
Formula Weight | 446.37 |
Percent Purity | 95% |
Color | White to Yellow |
Quantity | 5g |
Chemical Name or Material | Cefuroxime sodium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'