missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cefazolin Sodium Salt, MP Biomedicals™
Click to view available options
Quantity:
100 mg
500 mg
1 g
Specifications
Type | Cefazolin Sodium Salt |
Physical Form | Powder |
Formula Weight | 476.48 |
Color | White |
Quantity | 1 g |
Chemical Name or Material | Cefazolin Sodium Salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'