missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDX2 Mouse, Clone: CDX2/1690, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 1045-MSM1-P1
Additional Details : Weight : 0.01000kg
Specifications
CDX2 | |
Nuclear | |
CDX2/1690 | |
Unconjugated | |
XP_011533178 | |
Mouse | |
40kDa | |
Protein A/G Purified | |
RUO | |
1045 | |
IgG2a κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
CDX2 | |
Recombinant human CDX2 protein fragment (aa150-249) | |
HT29 cells or Colon Carcinoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |