missing translation for 'onlineSavingsMsg'
Learn More

CDPPB, Tocris Bioscience™

Positive allosteric modulator at mGlu5

Brand:  Tocris (Bio-Techne) 323510

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13462614

  • £162.36 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Brain penetrant, selective positive allosteric modulator at the mGlu5 receptor (EC50 values are 10 and 20 nM for human and rat receptors respectively). Antipsychotic; reverses amphetamine-induced locomotor activity and amphetamine-induced deficits in prepulse inhibition in rats.
Specifications

Chemical Identifiers

781652-57-1
364.408
cdppb, 3-cyano-n-1,3-diphenyl-1h-pyrazol-5-yl benzamide, mglur5 ligand, cdppb, d0d4hg, d0m5hy, 3-cyano-n-2,5-di phenyl pyrazol-3-yl benzamide, 3-cyano-n-1,3-diphenyl-1h-pyrazol-5-yl benzamide, 3-cyano-n-2,5-diphenyl-2h-pyrazol-3-yl-benzamide
3-cyano-N-(2,5-diphenylpyrazol-3-yl)benzamide
C23H16N4O
BKUIZWILNWHFHD-UHFFFAOYSA-N
11245456
C1=CC=C(C=C1)C2=NN(C(=C2)NC(=O)C3=CC=CC(=C3)C#N)C4=CC=CC=C4

Specifications

CDPPB
781652-57-1
cdppb, 3-cyano-n-1,3-diphenyl-1h-pyrazol-5-yl benzamide, mglur5 ligand, cdppb, d0d4hg, d0m5hy, 3-cyano-n-2,5-di phenyl pyrazol-3-yl benzamide, 3-cyano-n-1,3-diphenyl-1h-pyrazol-5-yl benzamide, 3-cyano-n-2,5-diphenyl-2h-pyrazol-3-yl-benzamide
C1=CC=C(C=C1)C2=NN(C(=C2)NC(=O)C3=CC=CC(=C3)C#N)C4=CC=CC=C4
364.408
364.4
10mg
C23H16N4O
BKUIZWILNWHFHD-UHFFFAOYSA-N
3-cyano-N-(2,5-diphenylpyrazol-3-yl)benzamide
11245456
>98%
Videos