missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ CDP-Star™ Substrate (0.25 mM Ready-To-Use)
CDP-Star™ Substrate (0.25 mM Ready-To-Use)
Brand: Molecular Probes™ T2147
Code : 50
Additional Details : Weight : 0.79000kg
Description
The CDP-Star™ Substrate is a chemiluminescent alkaline phosphatase substrate, ready-to-use for protein or nucleic acid blotting on nitrocellulose membranes. This substrate can also be used in solution-based assays.This CDP-Star™ chemiluminescent substrate for alkaline phosphatase let you detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed, and ease.
This versatile chemiluminescent substrate exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting. This substrate can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays. Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours.
For Research Use Only. Not for use in diagnostics procedures.
Specifications
250 mL | |
Bottle | |
CDP-Star™, NovaBright™ | |
Store the product at 4°C. |
Liquid | |
Southern, Northern, and Western Blot | |
Alkaline Phosphatase | |
Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.