missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CDK7/Cyclin H/MNAT1, His Tag Recombinant Protein
Human CDK7/Cyclin H/MNAT1, His Tag Recombinant Protein
£432.00 - £6870.00
Specifications
Gene Alias | CDK7/Cyclin H/MNAT1 |
---|---|
Species | Human |
Protein Tag | His Tag |
Expression System | Baculovirus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10046842
|
Invitrogen
PV4186 |
1mg |
£6870.00
1mg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15474603
|
Invitrogen
PR6749B |
100μg |
£1576.00
100µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11553597
|
Invitrogen
PV3868 |
10μg |
£432.00
10µg |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The trimeric complex of CDK7, cyclin H and MNAT1, functions as a Cdk-activating kinase (CAK). It is an essential component of the transcription factor TFIIH, that is involved in transcription initiation and DNA repair. This complex is thought to serve as a direct link between the regulation of transcription and the cell cycle.Specifications
CDK7/Cyclin H/MNAT1 | |
Human | |
His Tag | |
Baculovirus |
For Research Use Only. Not for use in diagnostic procedures.