missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CDK2/Cyclin A, His Tag Recombinant Protein
Human CDK2/Cyclin A, His Tag Recombinant Protein
£437.00 - £6900.00
Specifications
Accession Number | NP_001789 I NP_001228 |
---|---|
For Use With (Application) | Kinase Assay |
Gene Alias | CDK2/Cyclin A |
KinaseFamily | CDK and CHK Kinase Family |
Species | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10094102
|
Invitrogen™
PR4448A |
1mg |
£6900.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15438403
|
Invitrogen™
PR4448B |
100 μg |
£1604.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11563407
|
Invitrogen™
PV3267 |
10μg |
£437.00
10µg |
Estimated Shipment: 01-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CDK2 is a catalytic subunit of the cyclin-dependent protein kinase complex, and is essential for cell cycle G1⁄S phase transition. This protein is coexpressed and copurified with CyclinA.Specifications
NP_001789 I NP_001228 | |
CDK2/Cyclin A | |
Human | |
His Tag | |
Ser⁄Thr Kinases: CMCG Group | |
Baculovirus | |
Recombinant, Full Length |
Kinase Assay | |
CDK and CHK Kinase Family | |
Recombinant | |
Storage Buffer: 20mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 2mM DTT, 20% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Cell Cycle & Proliferation, Signal Transduction |
For Research Use Only. Not for use in diagnostic procedures.