missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CDK2/Cyclin A, His Tag Recombinant Protein
Human CDK2/Cyclin A, His Tag Recombinant Protein
Brand: Invitrogen™ PV3267
Code : 50
Additional Details : Weight : 3.36000kg
Description
CDK2 is a catalytic subunit of the cyclin-dependent protein kinase complex, and is essential for cell cycle G1⁄S phase transition. This protein is coexpressed and copurified with CyclinA.Specifications
NP_001789 I NP_001228 | |
1017, 890 | |
10μg | |
CDK and CHK Kinase Family | |
Recombinant | |
Storage Buffer: 20mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 2mM DTT, 20% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: CMCG Group | |
Z'-LYTE™, Omnia™ Bulk Substrate | |
cyclin-dependent kinase 2 | |
Kinases & Inhibitors | |
Serine/Threonine Kinases |
Kinase Assay | |
38.5⁄52.8 kDa | |
CDK2/Cyclin A | |
Human | |
His Tag | |
Biochemical binding/kinetic, Biochemical phospho-specific activity (no antibody) | |
Dry Ice | |
CDK2 | |
Baculovirus | |
Recombinant, Full Length | |
Cell Cycle & Proliferation, Signal Transduction |
For Research Use Only. Not for use in diagnostic procedures.