missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDK16 (PCTK1)/cyclin Y Recombinant Human Protein

CDK16 (PCTK1)/cyclin Y belongs to the cdc2/cdkx subfamily of the ser/thr family of protein kinases. Recombinant human CDK16 (107-end) and Cyclin Y (2-end) were co-expressed by baculovirus in Sf9 insect cells using N-terminal GST tags.

£431.00 - £6530.00

Specifications

For Use With (Application) Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
Formulation 50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol
Molecular Weight (g/mol) 71.1/65.5kDa
Storage Requirements Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Protein Length aa 108-end, 2-end
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15429013
View Documents Promotion Details
Invitrogen™
PV6379
10μg
£431.00
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 30-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15439013
View Documents Promotion Details
Invitrogen™
PV6380
100μg
£1460.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 08-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15449013
View Documents Promotion Details
Invitrogen™
PV6381
1mg
£6530.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 08-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CDK16 plays a role in signal transduction cascades in terminally differentiated cells, in exocytosis, and in transport of secretory cargo from the endoplasmic reticulum. CDK16 is ubiquitously expressed with the highest levels detected in the brain and testis. CDK16 also contained the gene coding for ubiquitin-activating enzyme UBE1 that previously shown to mapped to Xp11.3.

  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications

Specifications

Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
71.1/65.5kDa
aa 108-end, 2-end
Recombinant
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Human
GST Tag
Videos
SDS
Documents

Documents

Product Certifications