missing translation for 'onlineSavingsMsg'
Learn More

CD99 Mouse anti-Human, FITC, Clone: 3B2/TA8, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0997-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15566466

  • £132.00 / 100 tests

Estimated Shipment date
from Supplier 02-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD99 (E2, MIC2) is a transmembrane glycoprotein that is involved in regulation of T cell addhesive properties and programmed cell death distinct from typical apoptosis course. CD99 roles are specific to certain stages of T cell differentiation such as corticothymocytes. CD99R isoform expression is restricted in the haematopoietic system to T, NK and myeloid cells.
Specifications

Specifications

CD99
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P14209
HBA71, MIC2, MIC2X, MIC2Y, MSK5X
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
3B2/TA8
FITC
CD99
Liquid
CD99
IgG2a, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
4267
Videos
Documents

For Research Use Only.