missing translation for 'onlineSavingsMsg'
Learn More

CD98, PE, clone: RL388, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0981-83

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15218629

  • £195.47 / 200µg


Estimated Shipment date
from Supplier 27-06-2018
Add to basket

Description

Description

Description: The RL388 monoclonal antibody reacts with mouse CD98, also known as 4F2 antigen and Ly-10. RL388 detects a disulfide-linked heterodimer complex (130 kDa) composed of a glycosylated heavy (86 kDa) and a nonglycosylated light (39 kDa) subunits. CD98 is expressed on the surface of activated lymphocytes and is involved in cell activation and calcium flux events.Applications Reported: The RL388 antibody has been reported for use in flow cytometric analysis.Applications Tested: The RL388 antibody has been tested by flow cytometric analysis of activated and resting mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD98
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P10852
4F2, 4F2HC, 4T2HC, CD98, CD98HC, MDU1, NACAE
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
RL388
PE
Slc3a2
Conjugated
Slc3a2
IgG2a, kappa
200μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
17254
Videos

For Research Use Only.