Learn More
CD95 (APO-1/Fas), clone: 15A7, eBioscience™
Mouse Monoclonal Antibody
Brand:  Affymetrix eBioscience 14-0951-81
Code : Z2
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15207127
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Disclaimers:
For Research Use Only.
Description
Description
The 15A7 monoclonal antibody reacts with mouse CD95 (Fas, Apo-1), a 40-50kDa member of the TNFR superfamily. CD95 is expressed by a broad range of hematopoietic and non-hematopoietic cells including monocytes, neutrophils, activated lymphocytes and fibroblasts. Interaction of CD95 on mature lymphocytes with its ligand (FasL) induces apoptosis and is thought to be important in peripheral tolerance. 15A7 reacts with mouse CD95 in BALB/c strain and a bit more weakly in C57Bl/6 strain. 15A7 does not induce apoptosis of Fas-expressing cells.
Specifications
Specifications
CD95 (APO-1/Fas)
15A7
Unconjugated
Purified
IgG1
RUO
Primary
Murine
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
50μg
Store at 2-8°C.