Learn More
CD95 (APO-1/Fas) Monoclonal Antibody (15A7), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 17-0951-80
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The 15A7 monoclonal antibody reacts with mouse CD95 (Fas, Apo-1), a 40-50 kDa member of the TNFR superfamily. CD95 is expressed by a broad range of hematopoietic and non-hematopoietic cells including monocytes, neutrophils, activated lymphocytes and fibroblasts. Interaction of CD95 on mature lymphocytes with its ligand (FasL) induces apoptosis and is thought to be important in peripheral tolerance. 15A7 reacts with mouse CD95 in BALB/c strain and a bit more weakly in C57Bl/6 strain. 15A7 does not induce apoptosis of Fas-expressing cells. Applications Reported: This 15A7 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 15A7 antibody has been tested by flow cytometric analysis of mouse thymocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD95 (APO-1/Fas) Monoclonal antibody specifically detects CD95 (APO-1/Fas) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD95 (APO-1/Fas) | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P25446 | |
FAS | |
Affinity chromatography | |
RUO | |
14102 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
15A7 | |
APC | |
FAS | |
AI196731; ALPS1A; APO1; APO-1; Apo1 antigen; Apo-1 antigen; APO-1 cell surface antigen; Apo-1 Fas; apoptosis antigen 1; apoptosis signaling receptor FAS; apoptosis-mediating surface antigen FAS; APT1; CD95; CD95 antigen; FAS; Fas (TNF receptor superfamily member 6); Fas (TNF receptor superfamily member); Fas (TNF receptor superfamily, member 6); Fas AMA; Fas antigen; Fas antigen (ATP1); Fas cell surface death receptor; Fas receptor; FAS1; FASLG receptor; FASTM; lpr; lymphoproliferation; OTTHUMP00000020045; OTTHUMP00000020046; OTTHUMP00000020051; OTTHUMP00000059646; TNF receptor superfamily member 6; TNFR6; Tnfrsf6; tumor necrosis factor receptor superfamily member 6; tumor necrosis factor receptor superfamily member 6; tumor necrosis factor receptor superfamily member 26; Tumor necrosis factor receptor superfamily, member 6 | |
Mouse | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
For Research Use Only.