missing translation for 'onlineSavingsMsg'
Learn More

CD95 (APO-1/Fas), Alexa Fluor 488, clone: 15A7, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-0951-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15312000

  • £62.07 / 25µg


EU Stock 1
Add to basket

Description

Description

Description: The 15A7 monoclonal antibody reacts with mouse CD95 (Fas, Apo-1), a 40-50kDa member of the TNFR superfamily. CD95 is expressed by a broad range of hematopoietic and non-hematopoietic cells including monocytes, neutrophils, activated lymphocytes and fibroblasts. Interaction of CD95 on mature lymphocytes with its ligand (FasL) induces apoptosis and is thought to be important in peripheral tolerance. 15A7 reacts with mouse CD95 in BALB/c strain and a bit more weakly in C57Bl/6 strain. 15A7 does not induce apoptosis of Fas-expressing cells.Applications Reported: This 15A7 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 15A7 antibody has been tested by flow cytometric analysis of mouse thymocytes. This can be used at less than or equal to 2 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD95 (APO-1/Fas)
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P25446
ALPS1A, APO-1, APT1, Apo-1 Fas, CD95, FAS1, FASTM, TNFRSF6
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
15A7
Alexa Fluor 488
Fas
Conjugated
Fas
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
14102
Videos

For Research Use Only.