missing translation for 'onlineSavingsMsg'
Learn More

CD95 (APO-1/Fas), clone: DX2, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0959-82

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15247127

  • £105.87 / 100µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The DX2 monoclonal antibody reacts with human CD95 (Fas, Apo-1), a 40-50kDa member of the TNFR superfamily. CD95 is expressed by a broad range of hematopoietic and non-hematopoietic cells including monocytes, neutrophils, lymphocytes and fibroblasts. Interaction of CD95 on mature lymphocytes with its ligand (FasL) induces apoptosis and is thought to be important in peripheral tolerance. DX2 does not block binding of EOS9.1, another antibody specific for human CD95.Applications Reported: The DX2 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining. DX2 has also been reported in in vitro studies of Fas-induced apoptosis. EOS9.1 is recommended for induction of apoptosis in vitro .Applications Tested: The DX2 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD95 (APO-1/Fas)
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P25445
ALPS1A, APO-1, APT1, Apo-1 Fas, CD95, FAS1, FASTM, TNFRSF6
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry
DX2
Unconjugated
FAS
Purified
FAS
IgG1, kappa
100μg
Store at 2-8°C.
Primary
355
Videos

For Research Use Only.