missing translation for 'onlineSavingsMsg'
Learn More

CD94, FITC, clone: 18d3, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0941-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15218159

  • £43.65 / 50µg


Estimated Shipment date
from Supplier 30-05-2018
Add to basket

Description

Description

Description: The 18d3 monoclonal antibody reacts with mouse CD94, a 70kDa type II transmembrane glycoprotein. CD94 belongs to the C-type lectin superfamily and is present as a heterodimer with NKG2 on the surface. CD94 is expressed by NK cells, a subset of T cells, and NKT cells and plays an important role in adhesion and activation of NK cell lineage. DBA/2J mice are naturally CD94-deficient and do not express cell-surface CD94/NKG2A receptors, even on neonatal NK cells.Applications Reported: The 18d3 antibody has been reported for use in flow cytometric analysis.Applications Tested: The 18d3 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD94
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
O54707
Cluster of Differentiation 94
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
18d3
FITC
Klrd1
Conjugated
Klrd1
IgG2a, kappa
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
16643
Videos

For Research Use Only.