missing translation for 'onlineSavingsMsg'
Learn More

CD94 Mouse anti-Human, FITC, Clone: DX22, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0949-41

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15596456

  • £58.00 / 25 tests

Estimated Shipment
04-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Natural killer (NK) cells are a distinct lineage of lymphocytes that mediate cytotoxic activity and secrete cytokines upon immune stimulation. Several genes of the C-type lectin superfamily, including members of the NKG2 family, are expressed by NK cells and may be involved in the regulation of NK cell function. KLRD1 (CD94) is an antigen preferentially expressed on NK cells and is classified as a type II membrane protein because it has an external C terminus. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications

Specifications

CD94
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q13241
Cluster of Differentiation 94
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
DX22
FITC
KLRD1
Liquid
KLRD1
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3824
Videos
Documents

For Research Use Only.