missing translation for 'onlineSavingsMsg'
Learn More

CD94, FITC, clone: DX22, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0949-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15506466

  • £111.93 / 100 tests


EU Stock 13
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The DX22 monoclonal antibody reacts with human CD94, a 70kDa type II transmembrane glycoprotein. CD94 belongs to the C-type lectin superfamily and is present as a heterodimer with NKG2 on the surface. CD94 is expressed by NK cells, a subset of gd T cells, and NKT cells and plays an important role in adhesion and activation of NK cell lineage.Applications Reported: The DX22 antibody has been reported for use in flow cytometric analysis.Applications Tested: This DX22 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD94
DX22
FITC
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
KLRD1, KP43
IgG1
RUO
Primary
Human
Videos