missing translation for 'onlineSavingsMsg'
Learn More

CD93 (AA4.1) Mouse anti-Human, PE, Clone: R3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0938-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15566836

  • £72.50 / 25 tests

Estimated Shipment date
from Supplier 02-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications Tested: This R3 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. The protein encoded by this gene is a cell-surface glycoprotein and type I membrane protein that was originally identified as a myeloid cell-specific marker. The encoded protein was once thought to be a receptor for C1q, but now is thought to instead be involved in intercellular adhesion and in the clearance of apoptotic cells. The intracellular cytoplasmic tail of this protein has been found to interact with moesin, a protein known to play a role in linking transmembrane proteins to the cytoskeleton and in the remodelling of the cytoskeleton.
Specifications

Specifications

CD93 (AA4.1)
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9NPY3
C1QR1, C1qR(P), C1qRP, CDw93, ECSM3, MXRA4, dJ737E23.1
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
R3
PE
CD93
Liquid
CD93
IgM
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
22918
Videos
Documents

For Research Use Only.