missing translation for 'onlineSavingsMsg'
Learn More

CD93 (AA4.1) Mouse anti-Human, PE, Clone: R3, eBioscience™

Mouse Monoclonal Antibody

£72.50 - £167.00

Specifications

Antigen CD93 (AA4.1)
Clone R3
Target Species Human
Conjugate PE
Isotype IgM
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15566836 View Documents Affymetrix eBioscience
12-0938-41
25 Tests £72.50
25 tests View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 14-05-2020
Please to purchase this item. Need a web account? Register with us today!
15576836 View Documents Affymetrix eBioscience
12-0938-42
100 Tests £167.00
100 tests View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 15-04-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Applications Tested: This R3 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. The protein encoded by this gene is a cell-surface glycoprotein and type I membrane protein that was originally identified as a myeloid cell-specific marker. The encoded protein was once thought to be a receptor for C1q, but now is thought to instead be involved in intercellular adhesion and in the clearance of apoptotic cells. The intracellular cytoplasmic tail of this protein has been found to interact with moesin, a protein known to play a role in linking transmembrane proteins to the cytoskeleton and in the remodelling of the cytoskeleton.
Specifications

Specifications

CD93 (AA4.1)
Human
IgM
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
R3
PE
Flow Cytometry
Primary
Monoclonal
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.