missing translation for 'onlineSavingsMsg'
Learn More

CD90 (Thy-1) Mouse anti-Human, PerCP-eFluor™ 710, Clone: eBio5E10 (5E10), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-0909-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15579156

  • £116.00 / 25 tests

Estimated Shipment
30-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The eBio5E10 monoclonal antibody reacts with human CD90, also known as Thy-1 (thymus cell antigen-1). CD90 is a 25-35kDa receptor expressed on thymocytes, CD34+ prothymocytes, hematopoietic stem cells, neurons, a small subset of human fetal liver cells, cord blood cells, and bone marrow cells. CD90 is expressed on a subset of immature, CD34+ cells and a distinct subset of mature CD34- cells that are CD3+CD4+. The CD90+CD34+ population is enriched for cells capable of long-term culture. CD90 is involved in regulation of adhesion and signal transduction by T cells.
Specifications

Specifications

CD90 (Thy-1)
eBio5E10 (5E10)
PerCP-eFluor 710
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Thy1
IgG1, kappa
RUO
Primary
Human
Videos
Documents

For Research Use Only.