Learn More
CD90 (Thy-1) Monoclonal Antibody (eBio5E10 (5E10)), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£118.00 - £286.00
Specifications
Antigen | CD90 (Thy-1) |
---|---|
Clone | eBio5E10 (5E10) |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15528446
|
Affymetrix eBioscience
25-0909-41 |
25 Tests |
£118.00
25 tests |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15538446
|
Affymetrix eBioscience
25-0909-42 |
100 Tests |
£286.00
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The eBio5E10 monoclonal antibody reacts with human CD90, also known as Thy-1 (thymus cell antigen-1). CD90 is a 25-35 kDa receptor expressed on thymocytes, CD34+ prothymocytes, hematopoietic stem cells, neurons, a small subset of human fetal liver cells, cord blood cells, and bone marrow cells. CD90 is expressed on a subset of immature, CD34+ cells and a distinct subset of mature CD34- cells that are CD3+CD4+. The CD90+CD34+ population is enriched for cells capable of long-term culture. CD90 is involved in regulation of adhesion and signal transduction by T cells. Applications Reported: This eBio5E10 (5E10) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio5E10 (5E10) antibody has been pre-titrated and tested by flow cytometric analysis of human erythroleukemia (HEL) cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.
CD90 (Thy-1) Monoclonal antibody specifically detects CD90 (Thy-1) in Human samples. It is validated for Flow Cytometry.Specifications
CD90 (Thy-1) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
CD7; CD90; CDw90; FLJ33325; membrane glycoprotein; T25; Thy 1.2; THY1; Thy-1; Thy-1 antigen; Thy-1 cell surface antigen; thy-1 glycoprotein; thy-1 membrane glycoprotein; Thy-1 protein; Thy-1 T-cell antigen; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta; Thymus cell surface antigen | |
Thy1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Thy1 |
eBio5E10 (5E10) | |
Flow Cytometry | |
PE-Cyanine7 | |
Mouse | |
Human | |
P04216 | |
7070 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.