missing translation for 'onlineSavingsMsg'
Learn More

CD90 (Thy-1), APC, clone: eBio5E10 (5E10), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-0909-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15547976

  • £86.64 / 25 tests


EU Stock 1
Add to basket

Description

Description

Description: The eBio5E10 monoclonal antibody reacts with human CD90, also known as Thy-1 (thymus cell antigen-1). CD90 is a 25-35 kD receptor expressed on thymocytes, CD34+ prothymocytes, hematopoietic stem cells, neurons, a small subset of human fetal liver cells, cord blood cells, and bone marrow cells. CD90 is expressed on a subset of immature, CD34+ cells and a distinct subset of mature CD34- cells that are CD3+CD4+. The CD90+CD34+ population is enriched for cells capable of long-term culture. CD90 is involved in regulation of adhesion and signal transduction by T cells.Applications Reported: This eBio5E10 (5E10) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBio5E10 (5E10) antibody has been pre-titrated and tested by flow cytometric analysis of human erythroleukemia (HEL) cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD90 (Thy-1)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P04216
CD90
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
eBio5E10 (5E10)
APC
THY1
Conjugated
THY1
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
7070
Videos

For Research Use Only.