missing translation for 'onlineSavingsMsg'
Learn More

CD90.2 (Thy-1.2), PerCP-eFluor™ 710, clone: 30-H12, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-0903-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15381270

  • £57.60 / 25µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

The 30-H12 monoclonal antibody reacts with mouse CD90.2, also known as Thy-1.2, a GPI-linked membrane molecule. CD90.2 is expressed by mouse thymocytes and mature T cells as well as neurons in CD90.2-expressing mouse strains. These strains include BALB/c, CBA, C3H, C57BL/6, C58/, SJL and others. Cells from CD90.1-expressing strains including PL and AKR do not stain with 30-H12. CD90 is involved in regulation of adhesion and signal transduction by T cells.
Specifications

Specifications

CD90.2 (Thy-1.2)
30-H12
PerCP-eFluor 710
Conjugated
IgG2b, kappa
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos

For Research Use Only.