missing translation for 'onlineSavingsMsg'
Learn More

CD90.2 (Thy-1.2), Functional Grade, clone: 30-H12, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 16-0903-81

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15248527

  • £55.86 / 50µg


Estimated Shipment
01-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 30-H12 monoclonal antibody reacts with mouse CD90.2, also known as Thy-1.2, a GPI-linked membrane molecule. CD90.2 is expressed by mouse thymocytes and mature T cells as well as neurons in CD90.2-expressing mouse strains. These strains include BALB/c, CBA, C3H, C57BL/6, C58/, SJL and others. Cells from CD90.1-expressing strains including PL and AKR do not stain with 30-H12. CD90 is involved in regulation of adhesion and signal transduction by T cells.Applications Reported: The 30-H12 antibody has been reported for use in flow cytometric analysis. It has also been reported in in vivo and in vitro depletion.Applications Tested: The 30-H12 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Endotoxin: Less than 0.001 ng/ug antibody as determined by the LAL assay.Storage and handling: Use in a sterile environment.Filtration: 0.2μm post-manufacturing filtered.Purity: Greater than 90%, as determined by SDS-PAGE.Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay.Aggregation: Less than 10%, as determined by HPLC.
Specifications

Specifications

CD90.2 (Thy-1.2)
30-H12
Unconjugated
Purified
Rat
50μg
Store at 2-8°C.
Monoclonal
Functional Assay
1mg/mL
aqueous buffer, no sodium azide
Thy1.2
IgG2b
RUO
Primary
Murine
Videos