missing translation for 'onlineSavingsMsg'
Learn More

CD90.2 (Thy-1.2), Biotin, clone: 53-2.1, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0902-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15209649

  • £37.33 / 50µg


Estimated Shipment date
from Supplier 25-05-2018
Add to basket

Description

Description

Description: The 53-2.1 monoclonal antibody reacts with mouse CD90.2 also known as Thy-1.2, a GPI-linked membrane molecule. CD90.2 is expressed by mouse thymocytes and mature T cells as well as neurons in CD90.2-expressing mouse strains. These strains include BALB/c, CBA, C3H, C57BL/6, C58/, SJL and others. Cells from CD90.1-expressing strains including PL and AKR do not stain with 53-2.1. CD90 is involved in regulation of adhesion and signal transduction by T cells.Applications Reported: The 53-2.1 antibody has been reported for use in flow cytometric analysis.Applications Tested: The 53-2.1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD90.2 (Thy-1.2)
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P01831
CD90, T25, Thy-1, Thy-1.2, Thy1.1, Thy1.2,Thy 1.2, thy-1 antigen, thy-1 membrane glycoprotein
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
53-2.1
Biotin
Thy1
Conjugated
Thy1
IgG2a, kappa
50μg
Store at 2-8°C. Do not freeze.
Primary
21838
Videos

For Research Use Only.