missing translation for 'onlineSavingsMsg'
Learn More

CD90.1 (Thy-1.1), FITC, clone: HIS51, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0900-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15228149

  • £13.18 / 50µg


EU Stock 3
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

The HIS51 monoclonal antibody reacts with rat CD90 and cross-reacts with mouse CD90.1, a GPI-linked membrane molecule. In the rat, CD90 is expressed by hematopoietic stem cells, immature B cells, thymocytes, recent thymic emigrants, neurons, inflammed endothelia and other cell types. In the CD90.1-expressing mouse strains, PL and AKR, CD90 is expressed by early hematopoietic cells in the bone marrow, thymocytes and mature T cells.
Specifications

Specifications

CD90.1 (Thy-1.1)
HIS51
FITC
Conjugated
Murine
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Thy1.1
IgG2a
RUO
Primary
Rat
Videos