missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90.1 (Thy-1.1), eFluor 450, clone: HIS51, eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 48-0900-82
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The HIS51 monoclonal antibody reacts with rat CD90 and cross-reacts with mouse CD90.1, a GPI-linked membrane molecule. In the rat, CD90 is expressed by hematopoietic stem cells, immature B cells, thymocytes, recent thymic emigrants, neurons, inflammed endothelia and other cell types. In the CD90.1-expressing mouse strains, PL and AKR, CD90 is expressed by early hematopoietic cells in the bone marrow, thymocytes and mature T cells.Applications Reported: This HIS51 antibody has been reported for use in flow cytometric analysis.Applications Tested: This HIS51 antibody has been tested by flow cytometric analysis of rat splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.Specifications
CD90.1 (Thy-1.1) | |
Monoclonal | |
0.2mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
P01830, P01831 | |
THY1, Thy-1 cell surface antigen, FLJ33325, Thy-1 T-cell antigen, Thy-1 membrane glycoprotein | |
Murine | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Rat |
Flow Cytometry | |
HIS51 | |
eFluor 450 | |
Thy1 | |
Conjugated | |
Thy1 | |
IgG2a, kappa | |
100μg | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
Primary | |
21838 |
For Research Use Only.