missing translation for 'onlineSavingsMsg'
Learn More

CD90.1 (Thy-1.1), Biotin, clone: HIS51, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0900-85

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15299639

  • £131.50 / 500µg


Estimated Shipment date
from Supplier 04-04-2018
Add to basket

Description

Description

Description: The HIS51 monoclonal antibody reacts with rat CD90 and cross-reacts with mouse CD90.1, a GPI-linked membrane molecule. In the rat, CD90 is expressed by hematopoietic stem cells, immature B cells, thymocytes, recent thymic emigrants, neurons, inflammed endothelia and other cell types. In the CD90.1-expressing mouse strains, PL and AKR, CD90 is expressed by early hematopoietic cells in the bone marrow, thymocytes and mature T cells.Applications Reported: The HIS51 antibody has been reported for use in flow cytometric analysis.Applications Tested: The HIS51 antibody has been tested by flow cytometric analysis of rat thymocytes and splenocytes. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD90.1 (Thy-1.1)
HIS51
Biotin
Conjugated
Murine
500μg
Store at 2-8°C. Do not freeze.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Thy1.1
IgG2a, kappa
RUO
Primary
Rat
Videos

For Research Use Only.