Learn More
CD90.1 (Thy-1.1) Monoclonal Antibody (HIS51), Biotin, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£48.39 - £174.00
Specifications
Antigen | CD90.1 (Thy-1.1) |
---|---|
Clone | HIS51 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15289639
|
Affymetrix eBioscience
13-0900-81 |
50 μg |
£48.39
50µg |
Estimated Shipment: 08-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15299639
|
Affymetrix eBioscience
13-0900-85 |
500 μg |
£174.00
500µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The HIS51 monoclonal antibody reacts with rat CD90 and cross-reacts with mouse CD90.1, a GPI-linked membrane molecule. In the rat, CD90 is expressed by hematopoietic stem cells, immature B cells, thymocytes, recent thymic emigrants, neurons, inflammed endothelia and other cell types. In the CD90.1-expressing mouse strains, PL and AKR, CD90 is expressed by early hematopoietic cells in the bone marrow, thymocytes and mature T cells. Applications Reported: The HIS51 antibody has been reported for use in flow cytometric analysis. Applications Tested: The HIS51 antibody has been tested by flow cytometric analysis of rat thymocytes and splenocytes. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. It is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. Thy 1 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain.Specifications
CD90.1 (Thy-1.1) | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
CD7; CD90; FLJ33325; T25; Thy 1.2; Thy1; Thy-1; Thy-1 antigen; Thy-1 cell surface antigen; thy-1 glycoprotein; thy-1 membrane glycoprotein; Thy-1 protein; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta; Thymus cell surface antigen | |
Thy1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Thy1 |
HIS51 | |
Flow Cytometry | |
Biotin | |
Mouse | |
Mouse, Rat | |
P01830, P01831 | |
21838, 24832 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.